Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03648315_mH
          See other ABCG3 GE Assays ›
          Gene Symbol:
          Abcg3
          Gene Name:
          ATP-binding cassette, subfamily G (WHITE), member 3
          Gene Aliases:
          RGD1565568
          Chromosome Location:
          Chr.14: 5971984 - 6053039 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03648315_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcg3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128311.1 NP_001121783.1 1239 81

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Transporter 3' Most Recommended Assay Rattus norvegicus

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121782 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P220563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119553 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3899 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P220563 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P220563 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121782 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P220563 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771187 - -
          Affymetrix Rat Gene 1.0 ST Array 10771185 - -
          Affymetrix Rat Gene 1.0 ST Array 10775293 - -
          Affymetrix Rat Gene 1.0 ST Array 10775284 - -
          Affymetrix Rat Gene 1.0 ST Array 10771180 - -
          Affymetrix Rat Gene 1.0 ST Array 10775289 - -
          Affymetrix Rat Gene 1.0 ST Array 10775297 - -
          Affymetrix Rat Gene 1.0 ST Array 10775299 - -
          Affymetrix Rat Gene 1.0 ST Array 10771181 - -
          Affymetrix Rat Gene 1.0 ST Array 10771183 - -
          Affymetrix Rat Gene 1.0 ST Array 10775294 - -
          Affymetrix Rat Gene 1.0 ST Array 10771186 - -
          Affymetrix Rat Gene 1.0 ST Array 10775292 - -
          Affymetrix Rat Gene 1.0 ST Array 10771184 - -
          Affymetrix Rat Gene 1.0 ST Array 10775285 - -
          Affymetrix Rat Gene 1.0 ST Array 10775296 - -
          Affymetrix Rat Gene 1.1 ST Array 10775283 - -
          Affymetrix Rat Gene 1.1 ST Array 10771179 - -
          Affymetrix Rat Gene 1.0 ST Array 10771179 - -
          Affymetrix Rat Gene 1.0 ST Array 10775283 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351902 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0