Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04244351_m1
          See other CR2 GE Assays ›
          Gene Symbol:
          Cr2
          Gene Name:
          complement component (3d/Epstein Barr virus) receptor 2
          Gene Aliases:
          Cd21
          Chromosome Location:
          Chr.13: 113890274 - 113927814 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04244351_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105989.2 NP_001099459.2 15 - 16 2902 62
          XM_006250476.2 XP_006250538.1 19 - 20 3517 62
          XM_006250477.2 XP_006250539.1 15 - 16 2971 62
          XM_008769847.1 XP_008768069.1 22 - 23 4102 62
          XM_008769848.1 XP_008768070.1 15 - 16 3388 62
          GenBank mRNA FQ234665.1 - 2 - 3 386 62

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          complement receptor mediated signaling pathway
          B cell differentiation
          B cell proliferation
          B cell activation
          complement binding
          DNA binding
          receptor activity
          complement receptor activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P508580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49064 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508580 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508580 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P508580 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508573 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508580 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19995 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770871 - -
          Affymetrix Rat Gene 1.0 ST Array 10770885 - -
          Affymetrix Rat Gene 1.0 ST Array 10770875 - -
          Affymetrix Rat Gene 1.0 ST Array 10770860 - -
          Affymetrix Rat Gene 1.0 ST Array 10770863 - -
          Affymetrix Rat Gene 1.0 ST Array 10770876 - -
          Affymetrix Rat Gene 1.0 ST Array 10770864 - -
          Affymetrix Rat Gene 1.0 ST Array 10770869 - -
          Affymetrix Rat Gene 1.0 ST Array 10770862 - -
          Affymetrix Rat Gene 1.0 ST Array 10770870 - -
          Affymetrix Rat Gene 1.0 ST Array 10770868 - -
          Affymetrix Rat Gene 1.0 ST Array 10770873 - -
          Affymetrix Rat Gene 1.0 ST Array 10770874 - -
          Affymetrix Rat Gene 1.0 ST Array 10770872 - -
          Affymetrix Rat Gene 1.0 ST Array 10770865 - -
          Affymetrix Rat Gene 1.0 ST Array 10770859 - -
          Affymetrix Rat Gene 1.0 ST Array 10770866 - -
          Affymetrix Rat Gene 1.0 ST Array 10770867 - -
          Affymetrix Rat Gene 1.1 ST Array 10770858 - -
          Rat Genome 230 2.0 Array 1381327_PM_a_at - -
          Rat Genome 230 2.0 Array 1392233_at - -
          Rat Genome 230 2.0 Array 1385700_at - -
          Rat Genome 230 2.0 Array 1381327_a_at - -
          Rat Genome 230 2.0 Array 1381328_PM_at - -
          Rat Genome 230 2.0 Array 1392233_PM_at - -
          Rat Genome 230 2.0 Array 1385700_PM_at - -
          Rat Genome 230 2.0 Array 1381328_at - -
          Affymetrix HT Rat Focus Array 1381328_PM_at - -
          Affymetrix HT Rat Focus Array 1381327_a_at - -
          Affymetrix HT Rat Focus Array 1381327_PM_a_at - -
          Affymetrix HT Rat Focus Array 1381328_at - -
          Affymetrix HT Rat Focus Array 1392233_at - -
          Affymetrix HT Rat Focus Array 1385700_PM_at - -
          Affymetrix HT Rat Focus Array 1385700_at - -
          Affymetrix HT Rat Focus Array 1392233_PM_at - -
          Rat Expression Set 230 1385700_PM_at - -
          Rat Expression Set 230 1381328_PM_at - -
          Rat Expression Set 230 1385700_at - -
          Rat Expression Set 230 1381328_at - -
          Rat Expression Set 230 1392233_at - -
          Rat Expression Set 230 1392233_PM_at - -
          Rat Expression Set 230 1381327_a_at - -
          Rat Expression Set 230 1381327_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770858 - -
          Rat Genome U34 Set rc_AI177373_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21921235 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356697 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline