Search
Search
| Gene Symbol: Frmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271054.1 | NP_001257983.1 | 5 - 6 | 615 | 74 | |
| XM_006240151.2 | XP_006240213.1 | 5 - 6 | 665 | 74 | ||
| XM_006240152.2 | XP_006240214.1 | 6 - 7 | 745 | 74 | ||
| XM_006240153.2 | XP_006240215.1 | 5 - 6 | 1089 | 74 | ||
| GenBank mRNA | AF441249.1 | - | 4 - 5 | 392 | 74 | |
Set Membership: |
|