Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04342922_g1
          See other RPS23 GE Assays ›
          Gene Symbol:
          Rps23
          Gene Name:
          ribosomal protein S23
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 19823234 - 19824804 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04342922_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_078617.3 NP_511172.1 4 424 114
          GenBank mRNA BC058134.1 - 4 428 6918484 114
          FQ221811.1 - 4 416 114
          FQ222488.1 - 4 414 114
          FQ222494.1 - 4 418 114
          FQ222496.1 - 4 415 114
          FQ222641.1 - 4 416 114
          FQ223677.1 - 4 416 114
          FQ223812.1 - 4 416 114
          FQ228659.1 - 4 416 114
          FQ228931.1 - 4 416 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps23

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          regulation of translation
          structural constituent of ribosome
          poly(A) RNA binding
          translation regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049904 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065502 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P031072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333329 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065502 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031072 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065502 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049904 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P565385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812418 - -
          Affymetrix Rat Gene 1.0 ST Array 10812417 - -
          Affymetrix Rat Gene 1.0 ST Array 10772656 - -
          Affymetrix Rat Gene 1.0 ST Array 10812421 - -
          Affymetrix Rat Gene 1.0 ST Array 10812419 - -
          Affymetrix Rat Gene 1.0 ST Array 10812420 - -
          Affymetrix Rat Gene 1.0 ST Array 10802129 - -
          Affymetrix Rat Gene 1.1 ST Array 10812416 - -
          Affymetrix Rat Gene 1.1 ST Array 10772655 - -
          Affymetrix Rat Gene 1.1 ST Array 10802128 - -
          Rat Genome 230 2.0 Array 1370242_PM_at - -
          Rat Genome 230 2.0 Array 1370242_at - -
          Affymetrix HT Rat Focus Array 1370242_PM_at - -
          Affymetrix HT Rat Focus Array 1370242_at - -
          Affymetrix Rat Clariom S Assay PSR0200000648.rn.1 TC0200000132.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000647.rn.1 TC0200000132.rn.1 RTA TC
          Rat Expression Set 230 1370242_PM_at - -
          Rat Expression Set 230 1370242_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812416 - -
          Affymetrix Rat Gene 1.0 ST Array 10772655 - -
          Affymetrix Rat Gene 1.0 ST Array 10802128 - -
          Rat Genome U34 Set rc_AI013853_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22273870 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369998 - -
          RatRef-12 Expression BeadChip ILMN_48426 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline