Search
Search
| Gene Symbol: Ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031329.2 | NP_112619.2 | 7 - 8 | 1578 | 80 | |
| XM_006231853.2 | XP_006231915.1 | 7 - 8 | 1574 | 80 | ||
| XM_006231854.2 | XP_006231916.1 | 7 - 8 | 1618 | 80 | ||
| XM_006231855.2 | XP_006231917.1 | 6 - 7 | 1498 | 80 | ||
| GenBank mRNA | BC062037.1 | - | 7 - 8 | 1578 | 5623744 | 80 |
Set Membership: |
|