Search
Search
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053747.2 | NP_446199.2 | 4 - 5 | 849 | 68 | |
| XM_006253551.2 | XP_006253613.1 | 4 - 5 | 897 | 68 | ||
| GenBank mRNA | BC072477.1 | - | 4 - 5 | 849 | 7103662 | 68 |
| FQ222353.1 | - | 4 - 5 | 876 | 68 | ||
Set Membership: |
|