Search
Search
| Gene Symbol: Apbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053957.1 | NP_446409.1 | 8 - 9 | 1030 | 85 | |
| XM_006254495.1 | XP_006254557.1 | 8 - 9 | 1030 | 85 | ||
| GenBank mRNA | BC097935.1 | - | 6 - 7 | 1398 | 85 | |
| Y13413.1 | - | 8 - 9 | 1030 | 85 | ||
Set Membership: |
|