Search
Search
| Gene Symbol: Prom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138857.1 | NP_620212.1 | 8 - 9 | 1161 | 76 | |
| XM_006234947.2 | XP_006235009.1 | 8 - 9 | 1960 | 76 | ||
| GenBank mRNA | AF486828.1 | - | 8 - 9 | 1161 | 76 | |
| AF508942.1 | - | 8 - 9 | 1143 | 76 | ||
| FQ229034.1 | - | 5 - 6 | 706 | 76 | ||
Set Membership: |
|