Search
Search
| Gene Symbol: Strada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182820.1 | NP_877972.1 | 9 - 10 | 1113 | 82 | |
| XM_006247588.2 | XP_006247650.1 | 11 - 12 | 1291 | 82 | ||
| XM_006247595.2 | XP_006247657.1 | 10 - 11 | 1261 | 82 | ||
| GenBank mRNA | AY327409.1 | - | 9 - 10 | 1113 | 82 | |
| BC081911.1 | - | 10 - 11 | 1197 | 7111907 | 82 | |
Set Membership: |
|