Search
Search

Cyp20a1| Gene Symbol: Cyp20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199401.1 | NP_955433.1 | 2 - 3 | 166 | 70 | |
| GenBank mRNA | BC061716.1 | - | 2 - 3 | 166 | 5598443 | 70 |
Set Membership: |
|