Search
Search
| Gene Symbol: Irx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030044.1 | NP_001025215.1 | 2 - 3 | 649 | 84 | |
| XM_006255197.2 | XP_006255259.1 | 2 - 3 | 432 | 84 | ||
| XM_008772318.1 | XP_008770540.1 | 2 - 3 | 421 | 84 | ||
| GenBank mRNA | DQ109812.1 | - | 2 - 3 | 649 | 84 | |
Set Membership: |
|