Search
Search
| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034199.1 | NP_001029371.1 | 2 - 3 | 841 | 66 | |
| GenBank mRNA | CO395871.1 | - | 1 - 2 | 169 | 66 | |
| S82649.1 | - | 775 | 66 | |||
Set Membership: |
|