Search
Search

| Gene Symbol: RDN37-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132207.1 | 1 | 3487 | 70 | ||
| Gene Symbol: RDN37-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132216.1 | 1 | 3487 | 70 | ||
| Gene Symbol: TAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184514.1 | NP_690845.1 | 1 | 251 | 70 | |
Set Membership: |
|