Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss00968807_g1
See other CAMK2G GE Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
-
Chromosome Location:
Chr.14: 76535095 - 76632381 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss00968807_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_214193.1
NP_999358.1
6 - 7
501
76
XM_005671076.3
XP_005671133.1
500
76
XM_005671077.3
XP_005671134.1
541
76
XM_005671078.3
XP_005671135.1
531
76
XM_005671080.3
XP_005671137.1
492
76
XM_005671081.3
XP_005671138.1
557
76
XM_005671082.2
XP_005671139.1
713
76
XM_005671083.3
XP_005671140.1
512
76
XM_005671085.3
XP_005671142.1
503
76
XM_005671086.3
XP_005671143.1
581
76
XM_005671087.3
XP_005671144.1
523
76
XM_005671088.3
XP_005671145.1
531
76
XM_013983319.2
XP_013838773.1
541
76
XM_013983321.2
XP_013838775.1
541
76
XM_013983322.2
XP_013838776.1
503
76
XM_021072108.1
XP_020927767.1
482
76
XM_021072109.1
XP_020927768.1
540
76
XM_021072110.1
XP_020927769.1
482
76
XM_021072111.1
XP_020927770.1
482
76
XM_021072112.1
XP_020927771.1
506
76
XM_021072113.1
XP_020927772.1
479
76
XM_021072114.1
XP_020927773.1
531
76
XM_021072115.1
XP_020927774.1
490
76
XM_021072117.1
XP_020927776.1
522
76
XM_021072118.1
XP_020927777.1
539
76
XM_021072119.1
XP_020927778.1
503
76
XM_021072120.1
XP_020927779.1
455
76
GenBank mRNA
AK400720.1
-
8 - 9
713
76
U72970.1
-
6 - 7
474
76
U72971.1
-
6 - 7
449
76
U72973.1
-
6 - 7
501
76
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Detect Genomic DNA
Recommended Assay
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline