Search
Search
| Gene Symbol: MEIS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098046.1 | XP_020953705.1 | 2063 | 83 | ||
| XM_021098049.1 | XP_020953708.1 | 1247 | 83 | |||
| XM_021098056.1 | XP_020953715.1 | 2063 | 83 | |||
| XM_021098058.1 | XP_020953717.1 | 1177 | 83 | |||
| XM_021098059.1 | XP_020953718.1 | 2063 | 83 | |||
| XM_021098065.1 | XP_020953724.1 | 1246 | 83 | |||
| XM_021098074.1 | XP_020953733.1 | 2063 | 83 | |||
| XM_021098082.1 | XP_020953741.1 | 1245 | 83 | |||
| XM_021098084.1 | XP_020953743.1 | 2040 | 83 | |||
| XM_021098093.1 | XP_020953752.1 | 1565 | 83 | |||
| GenBank mRNA | AK234783.1 | - | 8 - 9 | 1067 | 83 | |
Set Membership: |
|