Search
Search
| Gene Symbol: LIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094649.1 | XP_020950308.1 | 2347 | 61 | ||
| GenBank mRNA | AK239228.1 | - | 22 - 23 | 2278 | 61 | |
| AK351665.1 | - | 22 - 23 | 2291 | 61 | ||
| AK392872.1 | - | 22 - 23 | 2341 | 61 | ||
Set Membership: |
|