Search
Search

LAMTOR1| Gene Symbol: LAMTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129591.3 | XP_003129639.1 | 570 | 61 | ||
| GenBank mRNA | AK236820.1 | - | 4 - 5 | 556 | 61 | |
| AK238446.1 | - | 4 - 5 | 521 | 61 | ||
| AK239877.1 | - | 4 - 5 | 516 | 61 | ||
| AK397802.1 | - | 4 - 5 | 531 | 61 | ||
| AY609478.1 | - | 4 - 5 | 498 | 61 | ||
Set Membership: |
|