Search
Search
| Gene Symbol: ALKBH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127080.6 | XP_003127128.3 | 230 | 78 | ||
| XM_021097221.1 | XP_020952880.1 | 230 | 78 | |||
| GenBank mRNA | AK345506.1 | - | 3 - 4 | 227 | 78 | |
Set Membership: |
|