Search
Search
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132619.5 | XP_003132667.1 | 1077 | 77 | ||
| XM_013982364.2 | XP_013837818.1 | 942 | 77 | |||
| XM_021070208.1 | XP_020925867.1 | 1159 | 77 | |||
| XM_021070209.1 | XP_020925868.1 | 1469 | 77 | |||
| XM_021070210.1 | XP_020925869.1 | 914 | 77 | |||
| XM_021070211.1 | XP_020925870.1 | 914 | 77 | |||
| XM_021070212.1 | XP_020925871.1 | 914 | 77 | |||
| XM_021070213.1 | XP_020925872.1 | 1005 | 77 | |||
| XM_021070214.1 | XP_020925873.1 | 917 | 77 | |||
| XM_021070215.1 | XP_020925874.1 | 709 | 77 | |||
| XM_021070216.1 | XP_020925875.1 | 517 | 77 | |||
Set Membership: |
|