Search
Search
| Gene Symbol: KDM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160090.1 | NP_001153562.1 | 12 - 13 | 1612 | 74 | |
| XM_021077210.1 | XP_020932869.1 | 1904 | 74 | |||
| XM_021077212.1 | XP_020932871.1 | 1745 | 74 | |||
| XM_021077215.1 | XP_020932874.1 | 1537 | 74 | |||
| XM_021077218.1 | XP_020932877.1 | 2478 | 74 | |||
| XM_021077224.1 | XP_020932883.1 | 102 | 74 | |||
| GenBank mRNA | EU661948.1 | - | 12 - 13 | 1612 | 74 | |
Set Membership: |
|