Search
Search
| Gene Symbol: RNF185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670858.3 | XP_005670915.1 | 412 | 84 | ||
| XM_005670859.3 | XP_005670916.1 | 418 | 84 | |||
| XM_013983075.2 | XP_013838529.2 | 524 | 84 | |||
| GenBank mRNA | AK232317.1 | - | 3 - 4 | 327 | 84 | |
| AK239947.1 | - | 3 - 4 | 320 | 84 | ||
| AK392989.1 | - | 3 - 4 | 396 | 84 | ||
Set Membership: |
|