Search
Search
| Gene Symbol: ABCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073710.1 | XP_020929369.1 | 3711 | 60 | ||
| GenBank mRNA | AF403247.1 | - | 2 - 3 | 154 | 60 | |
| DQ530510.1 | - | 5 - 6 | 735 | 60 | ||
Set Membership: |
|