Search
Search
| Gene Symbol: KPNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925212.7 | XP_001925247.4 | 1766 | 75 | ||
| XM_005654392.3 | XP_005654449.1 | 1194 | 75 | |||
| XM_005654393.3 | XP_005654450.1 | 1185 | 75 | |||
| GenBank mRNA | AF421152.1 | - | 3 - 4 | 351 | 75 | |
Set Membership: |
|