Search
Search
| Gene Symbol: BAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928147.4 | XP_001928182.1 | 640 | 64 | ||
| XM_013977773.2 | XP_013833227.1 | 915 | 64 | |||
| GenBank mRNA | AJ001204.1 | - | 2 - 3 | 202 | 64 | |
Set Membership: |
|