Search
Search
| Gene Symbol: ENPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087933.1 | XP_020943592.1 | 1647 | 68 | ||
| XM_021087944.1 | XP_020943603.1 | 1647 | 68 | |||
| GenBank mRNA | AK230988.1 | - | 5 - 6 | 434 | 68 | |
| DQ846654.1 | - | 16 - 17 | 1487 | 68 | ||
Set Membership: |
|