Search
Search
| Gene Symbol: GPAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005671462.3 | XP_005671519.2 | 5245 | 64 | ||
| GenBank mRNA | AK348444.1 | - | 16 - 17 | 2152 | 64 | |
| AY284842.1 | - | 2 - 3 | 319 | 64 | ||
Set Membership: |
|