Search
Search
| Gene Symbol: TBX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315722.1 | NP_001302651.1 | 6 | 1257 | 74 | |
| GenBank mRNA | AB183297.1 | - | 6 | 1257 | 74 | |
| AY435216.1 | - | 1 | 152 | 74 | ||
Set Membership: |
|