Search
Search
| Gene Symbol: MTHFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924329.5 | XP_001924364.2 | 3105 | 63 | ||
| XM_021099913.1 | XP_020955572.1 | 3250 | 63 | |||
| GenBank mRNA | AK238130.1 | - | 14 - 15 | 1512 | 63 | |
| AK239133.1 | - | 12 - 13 | 1278 | 63 | ||
| AK346354.1 | - | 27 - 28 | 3063 | 63 | ||
| AK393203.1 | - | 18 - 19 | 1898 | 63 | ||
| AY550929.1 | - | 4 - 5 | 374 | 63 | ||
Set Membership: |
|