Search
Search
| Gene Symbol: PROS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070736.1 | XP_020926395.1 | 98 | |||
| GenBank mRNA | AK236198.1 | - | 15 - 16 | 98 | ||
| L31379.1 | - | 13 - 14 | 1783 | 98 | ||
Set Membership: |
|