Search
Search
| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005652369.3 | XP_005652426.1 | 1104 | 79 | ||
| GenBank mRNA | AK233033.1 | - | 4 - 5 | 660 | 79 | |
| X56021.1 | - | 574 | 79 | |||
Set Membership: |
|