Search
Search
| Gene Symbol: AXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128458.1 | NP_001121930.1 | 12 | 1544 | 108 | |
| GenBank mRNA | AB364504.1 | - | 12 | 1544 | 108 | |
| AK395205.1 | - | 11 | 1481 | 108 | ||
Set Membership: |
|