Search
Search
| Gene Symbol: CYP7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230660.1 | - | 2 | 888 | 119 | |
| AK230789.1 | - | 7 | 1456 | 119 | ||
| AK230868.1 | - | 8 | 1564 | 119 | ||
| AK389629.1 | - | 7 | 1458 | 119 | ||
Set Membership: |
|