Search
Search
| Gene Symbol: NR3C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008481.1 | NP_001008481.1 | 8 | 2552 | 74 | |
| XM_005652952.3 | XP_005653009.2 | 2659 | 74 | |||
| XM_005652953.3 | XP_005653010.2 | 2323 | 74 | |||
| XM_005652954.3 | XP_005653011.2 | 2343 | 74 | |||
| XM_005652955.3 | XP_005653012.2 | 2579 | 74 | |||
| XM_005652956.3 | XP_005653013.2 | 2423 | 74 | |||
| XM_005652957.3 | XP_005653014.2 | 2766 | 74 | |||
| XM_013991680.2 | XP_013847134.2 | 2456 | 74 | |||
| XM_021080376.1 | XP_020936035.1 | 2414 | 74 | |||
| XM_021080385.1 | XP_020936044.1 | 2383 | 74 | |||
| XM_021080403.1 | XP_020936062.1 | 2203 | 74 | |||
| XM_021080408.1 | XP_020936067.1 | 2398 | 74 | |||
| XM_021080412.1 | XP_020936071.1 | 2453 | 74 | |||
| GenBank mRNA | AF141371.1 | - | 7 | 2046 | 74 | |
| AJ296022.1 | - | 7 | 2046 | 74 | ||
| AK343650.1 | - | 2 | 197 | 74 | ||
| AY779185.1 | - | 8 | 2552 | 74 | ||
| KC329457.1 | - | 7 | 2172 | 74 | ||
| U88894.1 | - | 3 | 393 | 74 | ||
Set Membership: |
|