Search
Search
| Gene Symbol: ID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037965.1 | NP_001033054.1 | 1 - 2 | 456 | 80 | |
| GenBank mRNA | AK231423.1 | - | 1 - 2 | 487 | 80 | |
| AK392405.1 | - | 1 - 2 | 485 | 80 | ||
| AY610248.1 | - | 1 - 2 | 464 | 80 | ||
| DQ116788.1 | - | 1 - 2 | 456 | 80 | ||
Set Membership: |
|