Search
Search
| Gene Symbol: TMSB4X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141991.2 | NP_001135463.2 | 2 | 149 | 107 | |
| GenBank mRNA | AK240144.1 | - | 2 | 131 | 107 | |
| AK392004.1 | - | 2 | 133 | 107 | ||
| AK400533.1 | - | 326 | 107 | |||
| DQ372079.1 | - | 1 | 64 | 107 | ||
Set Membership: |
|