Search
Search
| Gene Symbol: GATA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213879.1 | NP_999044.1 | 3 | 1052 | 93 | |
| XM_021068525.1 | XP_020924184.1 | 4077 | 93 | |||
| XM_021068526.1 | XP_020924185.1 | 987 | 93 | |||
| GenBank mRNA | AY251012.1 | - | 3 | 1052 | 93 | |
Set Membership: |
|