Search
Search
| Gene Symbol: ACOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101028.1 | NP_001094498.1 | 13 - 14 | 1950 | 62 | |
| XM_021066015.1 | XP_020921674.1 | 2311 | 62 | |||
| XM_021066016.1 | XP_020921675.1 | 1986 | 62 | |||
| XM_021066017.1 | XP_020921676.1 | 1945 | 62 | |||
| XM_021066018.1 | XP_020921677.1 | 2172 | 62 | |||
| XM_021066019.1 | XP_020921678.1 | 2038 | 62 | |||
| XM_021066020.1 | XP_020921679.1 | 4822 | 62 | |||
| GenBank mRNA | AK232470.1 | - | 13 - 14 | 2003 | 62 | |
| AK391508.1 | - | 9 - 10 | 1339 | 62 | ||
| AK391576.1 | - | 13 - 14 | 2035 | 62 | ||
| AK393248.1 | - | 13 - 14 | 2296 | 62 | ||
| AK394237.1 | - | 13 - 14 | 2008 | 62 | ||
| AK400728.1 | - | 14 - 15 | 2037 | 62 | ||
| AY609678.1 | - | 11 - 12 | 1604 | 62 | ||
| DQ842227.1 | - | 13 - 14 | 1950 | 62 | ||
| JQ087329.1 | - | 13 - 14 | 1948 | 62 | ||
| JQ087330.1 | - | 13 - 14 | 1948 | 62 | ||
Set Membership: |
|