Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03387661_u1
See other CHRM3 GE Assays ›
Gene Symbol:
CHRM3
Gene Name:
cholinergic receptor muscarinic 3
Gene Aliases:
-
Chromosome Location:
Chr.14: 51818625 - 52363662 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03387661_u1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CHRM3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001123098.1
NP_001116570.1
1
1542
108
XM_021071789.1
XP_020927448.1
5510
108
XM_021071790.1
XP_020927449.1
7795
108
XM_021071791.1
XP_020927450.1
5517
108
XM_021071792.1
XP_020927451.1
2642
108
XM_021071793.1
XP_020927452.1
4629
108
XM_021071794.1
XP_020927453.1
4636
108
XM_021071795.1
XP_020927454.1
2455
108
XM_021071796.1
XP_020927455.1
4013
108
XM_021071797.1
XP_020927456.1
4010
108
XM_021071798.1
XP_020927457.1
5387
108
XM_021071799.1
XP_020927458.1
1872
108
XM_021071800.1
XP_020927459.1
5493
108
XM_021071801.1
XP_020927460.1
5413
108
XM_021071802.1
XP_020927461.1
1904
108
XM_021071803.1
XP_020927462.1
3913
108
XM_021071804.1
XP_020927463.1
5307
108
XM_021071806.1
XP_020927465.1
5311
108
XM_021071807.1
XP_020927466.1
5452
108
XM_021071808.1
XP_020927467.1
5694
108
XM_021071809.1
XP_020927468.1
5244
108
XM_021071810.1
XP_020927469.1
5248
108
XM_021071811.1
XP_020927470.1
3807
108
XM_021071812.1
XP_020927471.1
2343
108
XM_021071813.1
XP_020927472.1
1688
108
XM_021071814.1
XP_020927473.1
2125
108
XM_021071815.1
XP_020927474.1
2012
108
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
G-protein coupled receptor
3' Most
Detect Genomic DNA
Recommended Assay
Sus scrofa
Inventoried
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline