Search
Search
| Gene Symbol: FOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214014.3 | NP_999179.3 | 2 | 1887 | 107 | |
| GenBank mRNA | AK234178.1 | - | 1 | 455 | 107 | |
| AK234453.1 | - | 3 | 871 | 107 | ||
| AY094061.1 | - | 2 | 1479 | 107 | ||
| DQ673620.1 | - | 1 | 172 | 107 | ||
| EF453379.1 | - | 2 | 1652 | 107 | ||
Set Membership: |
|