Search
Search
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044533.1 | NP_001037998.1 | 5 - 6 | 652 | 68 | |
| XM_021072183.1 | XP_020927842.1 | 659 | 68 | |||
| XM_021072184.1 | XP_020927843.1 | 651 | 68 | |||
| GenBank mRNA | AJ891123.1 | - | 5 - 6 | 652 | 68 | |
| AJ920266.1 | - | 5 - 6 | 597 | 68 | ||
| AJ920267.1 | - | 5 - 6 | 600 | 68 | ||
| AJ920268.1 | - | 5 - 6 | 588 | 68 | ||
| AJ920269.1 | - | 5 - 6 | 588 | 68 | ||
| AK398282.1 | - | 5 - 6 | 635 | 68 | ||
Set Membership: |
|