Search
Search

POMGNT1| Gene Symbol: POMGNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078669.1 | NP_001072137.1 | 14 - 15 | 1327 | 66 | |
| XM_005665452.3 | XP_005665509.1 | 1446 | 66 | |||
| XM_013999419.2 | XP_013854873.1 | 1519 | 66 | |||
| XM_021093615.1 | XP_020949274.1 | 1628 | 66 | |||
| GenBank mRNA | AB251343.1 | - | 14 - 15 | 1327 | 66 | |
| AK235556.1 | - | 6 - 7 | 354 | 66 | ||
| AK349483.1 | - | 15 - 16 | 1356 | 66 | ||
| AK391673.1 | - | 5 - 6 | 286 | 66 | ||
| AK393868.1 | - | 12 - 13 | 1058 | 66 | ||
Set Membership: |
|