Search
Search
| Gene Symbol: TRPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097459.1 | NP_001090928.1 | 7 - 8 | 2101 | 62 | |
| XM_005673829.3 | XP_005673886.1 | 3089 | 62 | |||
| GenBank mRNA | EF101315.1 | - | 7 - 8 | 2101 | 62 | |
Set Membership: |
|