Search
Search
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097517.1 | NP_001090986.1 | 4 - 5 | 765 | 82 | |
| XM_021068172.1 | XP_020923831.1 | 664 | 82 | |||
| GenBank mRNA | AK234353.1 | - | 4 - 5 | 765 | 82 | |
| EF486868.1 | - | 4 - 5 | 765 | 82 | ||
Set Membership: |
|