Search
Search
| Gene Symbol: IKBKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099935.1 | NP_001093405.1 | 14 - 15 | 1579 | 98 | |
| XM_021077168.1 | XP_020932827.1 | 1904 | 98 | |||
| XM_021077169.1 | XP_020932828.1 | 1827 | 98 | |||
| XM_021077170.1 | XP_020932829.1 | 1738 | 98 | |||
| XM_021077171.1 | XP_020932830.1 | 1866 | 98 | |||
| XM_021077172.1 | XP_020932831.1 | 1904 | 98 | |||
| GenBank mRNA | AB271927.1 | - | 14 - 15 | 1579 | 98 | |
| AK231306.1 | - | 15 - 16 | 1684 | 98 | ||
Set Membership: |
|