Search
Search

SLA-DOB| Gene Symbol: SLA-DOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114064.2 | NP_001107536.2 | 3 - 4 | 697 | 60 | |
| XM_021097879.1 | XP_020953538.1 | 980 | 60 | |||
| XM_021097880.1 | XP_020953539.1 | 906 | 60 | |||
| GenBank mRNA | AK236966.1 | - | 3 - 4 | 694 | 60 | |
| AK397732.1 | - | 3 - 4 | 700 | 60 | ||
| DQ333334.1 | - | 3 - 4 | 697 | 60 | ||
Set Membership: |
|