Search
Search
| Gene Symbol: PSMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213983.1 | NP_999148.1 | 3 - 4 | 315 | 75 | |
| XM_005668681.3 | XP_005668738.1 | 398 | 75 | |||
| XM_013980839.2 | XP_013836293.1 | 447 | 75 | |||
| XM_021066208.1 | XP_020921867.1 | 400 | 75 | |||
| XM_021066209.1 | XP_020921868.1 | 451 | 75 | |||
| XM_021066210.1 | XP_020921869.1 | 264 | 75 | |||
| XM_021066211.1 | XP_020921870.1 | 275 | 75 | |||
| GenBank mRNA | AK234194.1 | - | 3 - 4 | 205 | 75 | |
| AK395258.1 | - | 3 - 4 | 227 | 75 | ||
| AK396074.1 | - | 3 - 4 | 191 | 75 | ||
| X89719.1 | - | 143 | 75 | |||
Set Membership: |
|