Search
Search
| Gene Symbol: RHBG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213996.1 | NP_999161.1 | 8 - 9 | 1302 | 108 | |
| XM_021088694.1 | XP_020944353.1 | 1206 | 108 | |||
| XM_021088695.1 | XP_020944354.1 | 1318 | 108 | |||
| GenBank mRNA | AK234293.1 | - | 9 - 10 | 1304 | 108 | |
| AK351223.1 | - | 8 - 9 | 1316 | 108 | ||
| AY013261.1 | - | 8 - 9 | 1302 | 108 | ||
Set Membership: |
|