Search
Search
| Gene Symbol: GSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214050.2 | NP_999215.1 | 2 - 3 | 217 | 84 | |
| GenBank mRNA | AF188838.3 | - | 2 - 3 | 220 | 84 | |
| AK235250.1 | - | 2 - 3 | 369 | 84 | ||
| AK391766.1 | - | 2 - 3 | 244 | 84 | ||
| AY609655.1 | - | 2 - 3 | 366 | 84 | ||
Set Membership: |
|