Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ss03391797_m1
          See other BPIFA1 GE Assays ›
          Gene Symbol:
          BPIFA1
          Gene Name:
          BPI fold containing family A member 1
          Gene Aliases:
          PLUNC, SPLUNC1
          Chromosome Location:
          Chr.17: 36754498 - 36761622 on Build Sscrofa11.1
          Species:
          Pig
          Species Specific ID (Flybase ID):
          -
          Assay ID Ss03391797_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BPIFA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005727.1 NP_001005727.1 2 - 3 300 93
          XM_005672834.1 XP_005672891.1 479 93
          XM_005672835.2 XP_005672892.1 375 93
          GenBank mRNA AK238135.1 - 3 - 4 357 93
          AK238161.1 - 3 - 4 378 93
          AK238256.1 - 3 - 4 357 93
          AK238296.1 - 3 - 4 357 93
          AK350801.1 - 2 - 3 344 93
          AK350937.1 - 3 - 4 358 93
          AK397439.1 - 3 - 4 357 93
          AK397446.1 - 3 - 4 357 93
          AK397447.1 - 3 - 4 358 93
          AK397448.1 - 3 - 4 357 93
          AK397451.1 - 3 - 4 357 93
          AK397460.1 - 3 - 4 357 93
          AK397478.1 - 3 - 4 360 93
          AK397484.1 - 3 - 4 357 93
          AK397490.1 - 3 - 4 479 93
          AK397496.1 - 3 - 4 383 93
          AK397506.1 - 3 - 4 357 93
          AK397531.1 - 3 - 4 382 93
          AK397535.1 - 3 - 4 357 93
          AK397538.1 - 3 - 4 357 93
          AK397555.1 - 3 - 4 363 93
          AK397556.1 - 3 - 4 357 93
          AK397570.1 - 3 - 4 386 93
          AK397606.1 - 3 - 4 357 93
          AK397619.1 - 3 - 4 361 93
          AK397620.1 - 3 - 4 362 93
          AK397909.1 - 3 - 4 359 93
          AK397956.1 - 3 - 4 363 93
          AK397957.1 - 3 - 4 359 93
          AK397999.1 - 3 - 4 363 93
          AK398049.1 - 3 - 4 364 93
          AK398104.1 - 3 - 4 359 93
          AK399196.1 - 3 - 4 357 93
          AK399197.1 - 3 - 4 388 93
          AK399199.1 - 3 - 4 360 93
          AK399201.1 - 3 - 4 357 93
          AK399203.1 - 3 - 4 357 93
          AK399205.1 - 3 - 4 357 93
          AK399208.1 - 3 - 4 361 93
          AK399209.1 - 3 - 4 362 93
          AK399210.1 - 3 - 4 361 93
          AK399218.1 - 3 - 4 361 93
          AK399221.1 - 3 - 4 357 93
          AK399225.1 - 3 - 4 367 93
          AK399227.1 - 3 - 4 373 93
          AK399228.1 - 3 - 4 361 93
          AK399233.1 - 3 - 4 357 93
          AK399234.1 - 3 - 4 362 93
          AK399236.1 - 3 - 4 357 93
          AK399238.1 - 3 - 4 357 93
          AK399241.1 - 3 - 4 357 93
          AK399242.1 - 3 - 4 357 93
          AK399245.1 - 3 - 4 357 93
          AK399248.1 - 3 - 4 357 93
          AK399252.1 - 3 - 4 363 93
          AK399256.1 - 3 - 4 359 93
          AK399257.1 - 3 - 4 361 93
          AK399259.1 - 3 - 4 357 93
          AK399260.1 - 3 - 4 365 93
          AK399261.1 - 3 - 4 357 93
          AK399334.1 - 3 - 4 359 93
          AK399340.1 - 3 - 4 360 93
          AK399348.1 - 3 - 4 361 93
          AK399350.1 - 3 - 4 359 93
          AK399351.1 - 3 - 4 367 93
          AY733063.1 - 2 - 3 300 93

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          antibacterial humoral response
          innate immune response
          regulation of liquid surface tension
          multicellular organismal water homeostasis
          negative regulation of single-species biofilm formation in or on host organism
          regulation of sodium ion transmembrane transport
          lipid binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline