Search
Search
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011727.1 | NP_001011727.1 | 4 - 5 | 537 | 72 | |
| GenBank mRNA | AB527064.1 | - | 4 - 5 | 445 | 72 | |
| AK234979.1 | - | 4 - 5 | 471 | 72 | ||
| AK239495.1 | - | 4 - 5 | 571 | 72 | ||
Set Membership: |
|